异戊酸乙酯 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: Ethyl isopentanoate, Isovaleric acid ethyl ester, 3-Methylbutyric acid ethyl ester, Isopropyl acetic acid ethyl ester, 3-Methylbutanoic acid ethyl ester

150480050 150481000
ACRO150480050EA 0 CNY
ACRO150480050 ACRO150481000
异戊酸乙酯 99%
异戊酸乙酯
分子式: C₇H₁₄O₂
分子量: 130.19 g/mol
沸点: 131…133 °C (760 mmHg)
溶点: –99 °C
密度: 0.863 g/cm³
闪点: 26 °C
存储条件: Ambient
MDL编号: MFCD00009203
CAS号: 108-64-5
EINECS: 203-602-3
UN号: 1993
ADR: 3 III
Merck Index: 13,03849

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3980 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 0.861 to 0.865


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR