2-噻吩甲酸肼 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Thiophenecarboxylic hydrazide, 2-Thiophenecarboxylic acid hydrazide, Thiophene-2-carbohydrazide
138850050
138850250
ACRO138850050EA
0
CNY
ACRO138850050
ACRO138850250
2-噻吩甲酸肼 97%
2-噻吩甲酸肼
分子式:
C₅H₆N₂OS 分子量: 142.18 g/mol 溶点: 136…139 °C |
MDL编号:
MFCD00005435 CAS号: 2361-27-5 EINECS: 219-107-0 |
产品技术规格
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 136 to 140 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...