2-噻吩甲酸 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Thiophene-2-carboxylic acid, 2-Thiophenecarboxylic acid
138841000
138840050
ACRO138841000EA
0
CNY
ACRO138841000
ACRO138840050
2-噻吩甲酸 99%
2-噻吩甲酸
分子式:
C₅H₄O₂S 分子量: 128.15 g/mol 沸点: 260 °C (760 mmHg) 溶点: 126…130 °C 存储条件: Ambient |
MDL编号:
MFCD00005437 CAS号: 527-72-0 EINECS: 208-423-4 Merck Index: 12,09491 |
产品技术规格
Appearance | White to cream to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 126 to 130 °C |
GC | ≥98.5 % |
Titration with NaOH | 98.5 to 101.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...