四甲基脲 99%

供应商: Thermo Fisher Scientific
Danger

138491000 138492500
ACRO138491000EA 0 CNY
ACRO138491000 ACRO138492500
四甲基脲 99%
四甲基脲
分子式: C₅H₁₂N₂O
沸点: 174…178 °C (760 mmHg)
溶点: –1 °C
密度: 0,9879 g/cm³
闪点: 75 °C
存储条件: Ambient
MDL编号: MFCD00008319
CAS号: 632-22-4
EINECS: 211-173-9
Merck Index: 12,09368

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4520 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR