2-吡啶甲醇 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 2-(羟甲基)吡啶, 吡啶-2-甲醇, ω-羟基-2-皮考啉

131990250 131991000
ACRO131990250EA 0 CNY
ACRO131990250 ACRO131991000
2-吡啶甲醇 98%
2-吡啶甲醇
分子式: C₆H₇NO
分子量: 109.13 g/mol
沸点: 223
溶点: 5 °C
密度: 1.13 g/cm³
闪点: 117 °C
存储条件: Refrigerator
MDL编号: MFCD00006348
CAS号: 586-98-1
EINECS: 209-592-7

现在订购

产品技术规格

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5410 to 1.5450 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR