油胺 ∼ 80-90% C18
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
9-十八烯胺
129540010
129540050
129541000
ACRO129540010EA
0
CNY
ACRO129540010
ACRO129540050
ACRO129541000
油胺 ∼ 80-90% C18
油胺
分子式:
C₁₈H₃₇N 分子量: 267.5 g/mol 沸点: 364 溶点: 8…18 °C 密度: 0.81 g/cm³ 闪点: 154 °C 存储条件: Ambient |
MDL编号:
MFCD00066507 CAS号: 112-90-3 EINECS: 204-015-5 UN号: 2735 ADR: 8 II |
产品技术规格
Appearance | Clear yellow Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.4585 to 1.4625 (20°C, 589 nm) |
Titration with HClO4 | ≥96.0 % (Primary amine) |
Additional info | dielectric strength: ≥35 kV |
Viscosity | 0.7 to 0.9 cSt (at 25°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...