二甲基砜 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
二甲砜
127780050
127785000
ACRO127780050EA
0
CNY
ACRO127780050
ACRO127785000
二甲基砜 98%
二甲基砜
分子式:
C₂H₆O₂S 分子量: 94.13 g/mol 沸点: 238 °C (760 mmHg) 溶点: 107…111 °C 闪点: 143 °C 存储条件: Refrigerator |
MDL编号:
MFCD00007566 CAS号: 67-71-0 EINECS: 200-665-9 Merck Index: 13,03284 |
产品技术规格
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 107 to 111 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...