N-(2-羟乙基)酞酰亚胺 99%

供应商: Thermo Fisher Scientific

Synonyms: N-羟乙基酞酰亚胺

124920250 124921000
ACRO124920250EA 0 CNY
ACRO124920250 ACRO124921000
N-(2-羟乙基)酞酰亚胺 99%
N-(2-羟乙基)酞酰亚胺
分子式: C₁₀H₉NO₃
分子量: 191.19 g/mol
溶点: 126…129 °C
存储条件: Ambient
MDL编号: MFCD00005903
CAS号: 3891-07-4
EINECS: 223-434-4

现在订购

产品技术规格

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 129 °C
HPLC ≥98.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR