二苯并噻吩 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 二苯噻吩

112540250 112541000
ACRO112540250EA 0 CNY
ACRO112540250 ACRO112541000
二苯并噻吩 98%
二苯并噻吩
分子式: C₁₂H₈S
分子量: 184.26 g/mol
沸点: 332…333 °C (760 mmHg)
溶点: 97…100 °C
闪点: 170 °C
存储条件: Ambient
MDL编号: MFCD00004969
CAS号: 132-65-0
EINECS: 205-072-9
UN号: 3077
ADR: 9 III

现在订购

产品技术规格

Appearance White to beige to grey Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 97 to 101 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR