N-(3-溴丙基)邻苯二甲酰亚胺 98%

供应商: Thermo Fisher Scientific

107390250 107391000
ACRO107390250EA 0 CNY
ACRO107390250 ACRO107391000
N-(3-溴丙基)邻苯二甲酰亚胺 98%
N-(3-溴丙基)邻苯二甲酰亚胺
分子式: C₁₁H₁₀BrNO₂
分子量: 268.11 g/mol
沸点: 433
溶点: 70…76 °C
MDL编号: MFCD00005904
CAS号: 5460-29-7
EINECS: 226-738-5

现在订购

产品技术规格

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 70 to 76 °C
HPLC ≥97.5 %
Iron (Fe) ≤100 ppm
Heavy metals (as Pb) ≤50 ppm
Nitrilotriacetic acid ≤0.1 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR