1-苯基-1,3-丁二酮 98%

供应商: Thermo Fisher Scientific

Synonyms: 苯甲酰丙酮

105690500 105692500
ACRO105690500EA 0 CNY
ACRO105690500 ACRO105692500
1-苯基-1,3-丁二酮 98%
1-苯基-1,3-丁二酮
分子式: C₆H₅C(O)CH₂C(O)CH₃
沸点: 262 °C (1013 hPa)
溶点: 54…58 °C
密度: 1,09 g/cm³
存储条件: Ambient
MDL编号: MFCD00008786
CAS号: 93-91-4
EINECS: 202-286-4

现在订购

产品技术规格

Appearance Yellow Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR