2-吡啶甲基胺 99%

供应商: Thermo Fisher Scientific
Danger

104070250 104071000
ACRO104070250EA 0 CNY
ACRO104070250 ACRO104071000
2-吡啶甲基胺 99%
2-吡啶甲基胺
分子式: C₆H₈N₂
沸点: 82…85 °C (12 mmHg)
溶点: –20 °C
密度: 1,04 g/cm³
闪点: 90 °C
存储条件: Ambient
MDL编号: MFCD00006360
CAS号: 3731-51-9
EINECS: 223-090-5
UN号: 2735
ADR: 8,II

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5430 to 1.5470 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤15 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR