(±)-Mandelic acid 99+%
供应商: Thermo Fisher Scientific
|
Danger
|
125310010
125311000
125312500
ACRO125310010EA
0
CNY
ACRO125310010
ACRO125311000
ACRO125312500
(±)-Mandelic acid 99+%
(±)-Mandelic acid
分子式:
C₈H₈O₃ 沸点: 322 °C (1013 hPa) 溶点: 118…122 °C 密度: 1,361 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00064250 CAS号: 90-64-2 EINECS: 210-277-1 |
产品技术规格
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 117 to 121 °C |
Titration with NaOH | ≥99.0 % |
Nitrate (NO3) | ≤0.001 % |
Phosphate (PO4) | ≤0.0003 % |
Aluminium (Al) | ≤0.001 % |
Arsenic (As) | ≤0.00002 % |
Cadmium (Cd) | ≤0.0005 % |
Calcium (Ca) | ≤0.001 % |
Chromium (Cr) | ≤0.0005 % |
Cobalt (Co) | ≤0.0005 % |
Copper (Cu) | ≤0.0005 % |
Iron (Fe) | ≤0.002 % |
Lead (Pb) | ≤0.0005 % |
Magnesium (Mg) | ≤0.0005 % |
Manganese (Mn) | ≤0.0005 % |
Nickel (Ni) | ≤0.0005 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.005 % |
Zinc (Zn) | ≤0.0005 % |
Heavy metals (as Pb) | ≤20 ppm |
Insoluble matter | ≤0.005 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...