DL-4-羟基-3-甲氧基扁桃酸 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 3-甲氧基-4-羟基扁桃酸, VMA, 香草扁桃酸, 4-羟基-3-甲氧基扁桃酸, 4-羟基-3-甲氧基苦杏仁酸

164900010 164900050
ACRO164900010EA 0 CNY
ACRO164900010 ACRO164900050
DL-4-羟基-3-甲氧基扁桃酸 99%
DL-4-羟基-3-甲氧基扁桃酸
分子式: C₉H₁₀O₅
分子量: 198.18 g/mol
沸点: 421
溶点: 131…134 °C (dec.)
存储条件: Refrigerator
MDL编号: MFCD00004235
CAS号: 55-10-7
EINECS: 200-224-0
Merck Index: 12,10070

现在订购

产品技术规格

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 131 to 134 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR