Semi-Micro Balances
供应商: METTLER TOLEDO
MS105 MS105DU MS205DU
METBMS105EA 0 CNY
METBMS105 METBMS105DU METBMS205DU
Semi-Micro Balances
天平
These MS Semi-Micro Balances offer strong performance and increased productivity thanks to a fast settling time and consistently reliable results.
  • Strong performance
  • Increased productivity
  • Fast settling time
  • Reliable results
  • ErgoDoors
  • Built-in date and time functions
The Swiss high-precision weighing cell HRT guarantees constant accuracy over the entire weighing range. Clever ErgoDoors make the weighing-in of samples as easy and convenient as possible. The built-in date and time function enables GLP/GMP compliant print outs, ensuring full control and traceability for your research.