Value Plastics® Adapter Fittings, Male Luer to Hose Barb, Straight, Nylon
供应商: MASTERFLEX PREMIUM
45505-00BLK 45505-04BLK
MFLB45505-00BLKEA 0 CNY
MFLB45505-00BLK MFLB45505-04BLK MFLA45505-06BLK MFLA45505-08BLK MFLA45505-19BLK MFLA45505-33BLK MFLA45505-02BLK MFLA45505-32BLK MFLA45505-22BLK MFLA45505-31BLK
Value Plastics® Adapter Fittings, Male Luer to Hose Barb, Straight, Nylon
接头
Ensure your process integrity with full material traceability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.