Value Plastics® Adapter Fittings, Luer to Threaded
供应商: MASTERFLEX PROPRIETARY
MFLA45505-78BLKEA 0 CNY
MFLA45505-78BLK MFLA45505-80BLK MFLA45505-88BLK MFLA45505-86BLK MFLA45502-66BLK MFLA45505-82BLK MFLA45505-76BLK MFLA45502-60BLK MFLA45505-84BLK
Value Plastics® Adapter Fittings, Luer to Threaded
接头
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Easily complete your luer fittings systems with these premium connectors. Included are some special configurations available only in polycarbonate. The snap luer lock ring is for use with male luer by wide-bore hose barb fittings. Nylon material has good chemical resistance and can be sterilized via autoclave, ethylene oxide, or gamma irradiation.