乙腈 ≥99.9% ACS for HPLC

供应商: Honeywell Chemicals
Danger

Synonyms: 甲基氰, ACN

AH015-4 AH015-4HC-1
HONCAH015-4EA 0 CNY
HONCAH015-4 HONCAH015-4HC-1
乙腈 ≥99.9% ACS for HPLC
乙腈
分子式: H₃CCN
沸点: 82 °C (1,013 hPa)
溶点: –46 °C
密度: 0,7822 g/cm³ (20 °C)
闪点: 6 °C
存储条件: Ambient
MDL编号: MFCD00001878
CAS号: 75-05-8
EINECS: 200-835-2
UN号: 1648
ADR: 3,II

现在订购

产品技术规格

Color (APHA) 10 max.
Water by Karl Fisher Titration 0.05 % max.
GC Analysis 99.9 % min.
HPLC Gradient @ 254 nm 0.005 AU max.
Refractive Index (20ºC) 1.3434 - 1.3446
Residue 3 mg/L max.
Titrable Acid 8 µeq/g max.
Titrable Base 0.6 µeq/g max.
UV Absorbance @ 200 nm 0.07 AU max.
UV Cutoff 190 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR