Union Fittings, Compression, Straight
供应商: MASTERFLEX PREMIUM
31406-00 31406-50 31412-00 31412-50
MFLB31406-00EA 0 CNY
MFLB31406-00 MFLB31406-50 MFLB31412-00 MFLB31412-50
Union Fittings, Compression, Straight
接头
Get a secure seal at higher pressures.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).