Masterflex® Transfer Tubing, Tygon® Tygon® S3™ E-3603, Avantor®
供应商: MASTERFLEX PROPRIETARY
06407-99
MFLA06407-70EA 0 CNY
MFLA06407-70 MFLA06407-71 MFLA06407-72 MFLA06407-73 MFLA06407-74 MFLA06407-75 MFLA06407-76 MFLA06407-77 MFLA06407-78 MFLA06407-79 MFLA06407-80 MFLA06407-81 MFLA06407-82 MFLA06407-83 MFLA06407-84 MFLA06407-85 MFLA06407-86 MFLA06407-87 MFLA06407-88 MFLA06407-89 MFLA06407-90 MFLA06407-91 MFLA06407-92 MFLA06407-93 MFLA06407-94 MFLA06407-95 MFLA06407-96 MFLA06407-98 MFLB06407-99
Masterflex® Transfer Tubing, Tygon® Tygon® S3™ E-3603, Avantor®
胶管 Transfer Tubing
Pliable formulation for sharp radius bending.
  • 配方:Tygon E-3603
  • 应用:一般实验室,食品与饮料,生物制药,分析仪器。
  • 特征:无-DEHP,无邻苯二甲酸酯。使用寿命长,耐化学性,无氧化性;可以弯曲成急弯半径曲线,配上接头使用简单。透明。
  • 认证:满足FDA要求;符合USP VI级和NSF 51要求。
  • 温度范围:–51至165°F (–46至74°C)
  • 消毒:高温高压或环氧乙烷。
This tubing is ideal for general laboratory use. It offers long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, and is nontoxic.
FDA compliant and made according to GMP.
Temperature range is -51 to 165 °F (-46 to 73 °C)
Can be sterilized with ethylene oxide or autoclave