Masterflex® Transfer Tubing, Versilon™ A-60-G
供应商: MASTERFLEX PREMIUM
06410-01 06410-02 06410-03 06410-04 06410-05 06410-06 06410-07 06410-08 06410-09 06410-10 06410-11 06410-13 06410-14 06410-15 06410-17 06410-18 06410-12 06410-16 06410-19
MFLB06410-01EA 0 CNY
MFLB06410-01 MFLB06410-02 MFLB06410-03 MFLB06410-04 MFLB06410-05 MFLB06410-06 MFLB06410-07 MFLB06410-08 MFLB06410-09 MFLB06410-10 MFLB06410-11 MFLB06410-13 MFLB06410-14 MFLB06410-15 MFLB06410-17 MFLB06410-18 MFLB06410-12 MFLB06410-16 MFLB06410-19
Masterflex® Transfer Tubing, Versilon™ A-60-G
胶管 Transfer Tubing
Resists chemicals and UV-light.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
应用:要求优秀的耐化学性,耐热,防臭氧和防紫外线(UV)的应用。
特征:绝缘,可热封,非老化,抗强酸强碱。不透明黑色。
认证:泵管依据GMP生产。
温度范围:–75 至 275°F (–60 至 135°C)
消毒:高温高压杀菌。