Noryl® Corrosion Resistant Chemical-Resistance Centrifugal Pumps
供应商: Avantor Fluid Handling
70840-15 70840-35
MFLS70840-15EA 0 CNY
MFLS70840-15 MFLS70840-35
Noryl® Corrosion Resistant Chemical-Resistance Centrifugal Pumps
Centrifugal Pumps
Chemical-resistant pump heads and impellers handle a variety of corrosive and caustic fluids over the entire pH range.
  • Ideal for circulation, transfer, and chemical feed applications
  • NEMA 56 C-face motors feature thermal overload protection
These Noryl® corrosion resistant chemical-resistance centrifugal pumps are ideal for circulation, transfer, and chemical feed applications. Pumps are constructed with a Noryl pump head and impeller, carbon/ceramic face seal, Noryl seal encasement, and Viton® O-ring.
Delivery information: Includes all pumps come with stainless steel plated hardware and conduit box for hardwiring.