Masterflex® 乙烯基泵管,Avantor®
供应商: MASTERFLEX PREMIUM
06405-01 06405-03 06405-04 06405-06 06405-07 06405-08 06405-09 06405-10 06405-12 06405-15 06405-16 06405-18 06405-19 06405-20 06405-31 06405-36 06405-02 06405-27 06405-25 06405-46
MFLB06405-01EA 0 CNY
MFLB06405-01 MFLB06405-03 MFLB06405-04 MFLB06405-06 MFLB06405-07 MFLB06405-08 MFLB06405-09 MFLB06405-10 MFLB06405-12 MFLB06405-15 MFLB06405-16 MFLB06405-18 MFLB06405-19 MFLB06405-20 MFLB06405-31 MFLB06405-36 MFLB06405-02 MFLB06405-27 MFLB06405-25 MFLB06405-46
Masterflex® 乙烯基泵管,Avantor®
胶管 Transfer Tubing
Ideal for general-purpose applications.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
应用:普通功能实验室使用。
特性:特征:良好的抗化学腐蚀性和抗研磨性,无嗅,无毒;透明。
认证:泵管依据GMP制造。
温度范围: –45 至 180°F (–43 至 82°C)。
消毒:环氧乙烷。