Blue-White® 单头化学品灌注隔膜泵
供应商: MASTERFLEX PREMIUM
76302-20 76302-30 76302-40 76302-00 76302-10 76302-15 76302-25 76302-35
MFLB76302-20EA 0 CNY
MFLB76302-20 MFLB76302-30 MFLB76302-40 MFLB76302-00 MFLB76302-10 MFLB76302-15 MFLB76302-25 MFLB76302-35
Blue-White® 单头化学品灌注隔膜泵
Diaphragm Pumps
Economical, chemically-compatible diaphragm metering pumps.
  • 长距离轻松灌注
  • 低冲数/分钟率延长了隔膜寿命
  • 过载保护、风扇冷却、罩极式齿轮电机 (1/35 hp) 为终身润滑
  • 密封 VALOX® 外壳可以耐受恶劣天气条件
该泵成套发货,包括脚踩阀/滤网,聚丙烯防虹吸阀,乙烯基入口管和聚乙烯出口阀,安装器具,和6英尺三线制电源线和插头。
Delivery information: Includes foot valve/strainer, polypropylene antisiphon valve, vinyl inlet and polyethylene outlet tubing, mounting hardware, and 6-ft, three-wire cord and plug.