Magnetic Drive Centrifugal Pumps with Open Drip Proof (ODP) Motor
供应商: MASTERFLEX PREMIUM
07004-32 07021-00 07021-01 07021-02 07021-03 07022-22 07023-12 07022-20 07023-05 07023-06
MFLB07004-32EA 0 CNY
MFLB07004-32 MFLB07021-00 MFLB07021-01 MFLB07021-02 MFLB07021-03 MFLB07022-22 MFLB07023-12 MFLB07022-20 MFLB07023-05 MFLB07023-06
Magnetic Drive Centrifugal Pumps with Open Drip Proof (ODP) Motor
Centrifugal Pumps
Polypropylene (PP)-encapsulated magnets offer excellent chemical resistance.
  • Easily disassemble pumps for cleaning
  • ODP pumps provide an economical option for use in chemical OEM and industrial applications
These magnetic drive centrifugal pumps with open drip proof (ODP) motors are excellent for applications that require chemical resistance. Pumps are constructed with a polypropylene (PP) body and impeller, Viton® O-ring, and PP-encapsulated ceramic spindle. Maximum temperature of 190˚F (88˚C).
60赫兹时,泵的流量和扬程为50赫兹时的5/6。
美国杜邦公司VITON系列产品。