VWR®, Refrigerated circulating baths, 240 V
供应商: VWR BRAND PRIVATE LABEL USA
89202-996 89203-024
VWRU89202-996EA 0 CNY
VWRU89202-996 VWRU89203-024
VWR®, Refrigerated circulating baths, 240 V
循环水浴 闭式循环系统 Cooling Circulators
VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40 to 200°C (depending on the controller).
  • Easy-to-view LCD display
  • Precise temperature control
Available in a wide range of sizes, with three controller options, these refrigerated circulating baths are ideal for many applications.
The large LCD display on the MX temperature controller is easily viewed across the lab. The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature Controller includes a full touch-screen display, and programmability (ten 100-step programs).
The Advanced Series Controllers offer Swivel 180™ technology, which allows you to rotate the controller independently from the bath. All heated circulating baths include LidDock™ lid stowing system and DuraTop™, the precision molded phenolic bath top, which remains cooler at high temperatures and resists laboratory chemicals.
Certifications: ETL listed. CE marked. MX Controller complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. Advanced Digital and Advanced Programmable comply with DIN 12876-1 Class III safety requirements for use with flammable liquids. cETLus compliant.
订购须知 Refrigerated Circulators include reservoir cover; bypass tubing; male inlet and outlet adapters for 9,5, 6,3 ,4,7 mm tubing. 1/4" NPT to M16 adapters also included on 50 Hz models.