Chemical resistant overalls, Tyvek® IsoClean®, model IC183
供应商: DUPONT PROTECTIVE APPAREL
D15429659 D15429666 D15429671 D15429681 D15429648
DUPOD15429659EA 0 CNY
DUPOD15429659 DUPOD15429666 DUPOD15429671 DUPOD15429681 DUPOD15429648
Chemical resistant overalls, Tyvek® IsoClean®, model IC183
Disposable Garments
DuPont™ Tyvek® IsoClean® unhooded coverall model IC 183 B WH DS. Clean-processed and gamma-sterilised. Bound internal seams. Tunnelled elastication at wrists and ankles.
  • Elasticated waist at back
  • Tyvek® covered elasticated thumb loops
  • Zipper closure
  • Storm flap
  • Aseptically folded
  • Clean-processed and sterilised by gamma-irradiation to SAL of 10⁻⁶
Suitable for use in GMP class A/B (ISO Class 5) clean rooms. Basis weight of 45 g/m².
Certifications: Certified according to regulation (EU) 2016/425; EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination); Chemical protective clothing, Category III, Type 5-B and 6-B.
Packaging: 25 per box, individually double packed. 2 polyethylene liners. Cardboard box.