Cleanroom gloves, latex, Kimtech Pure* G5
供应商: KIMBERLY CLARK
56852 56853 56854 56855
KIMB56852EA 0 CNY
KIMB56852 KIMB56853 KIMB56854 KIMB56855
Cleanroom gloves, latex, Kimtech Pure* G5
手套
Sterile gloves made of natural rubber latex.
  • Beaded cuffs
  • Textured palm and palm side on fingertips
  • Gamma irradiated
Certifications: Validated to a Sterility Assurance Level (SAL) of 10⁻⁶ according to ANSI/AAMI/ISO 11137.
Packaging: 1 pair/coated paper wallet, 1 wallet/ pouch, 20 pouches/ double bag, 10 double bags per lined case; 200 pairs per case.