These reliable, easy-to-wear gloves are more economical than standard nitrile gloves and are ideal for a wide variety of tasks. The lavender nitrile gloves deliver enhanced dexterity thanks to textured fingertips.
- Exceptional value: 150% more gloves per case (versus standard nitrile gloves)
- Latex-free, powder free
- Beaded cuffs for easier donning
- Static dissipative in use
- Superior quality, AQL 1,5 (exceeds the ASTM D 5151 standard for critical defects or pinholes)
- Less waste thanks to the one-at-a-time dispenser box
- Tested for use with 12 chemotherapy drugs and four hormones (ASTM D6978-05)
The gloves come in an innovative dispenser box, which incorporates two separate openings. The first, smaller opening is used when the box is full and helps control dispensing to lessen waste from dropped gloves or multiple dispensing. When the box is half empty, the second, larger opening allows easier access to the gloves. They have an AQL of 1,5 for pinholes and can even be recycled with the Kimberly-Clark Professional RightCycle program, the first large-scale recycling program for nonhazardous lab and cleanroom waste. It can be an important piece for creating a more efficient, responsible workplace.