Wipes for general applications, WYPALL* L30
供应商: KIMBERLY CLARK
83030 05812
KIMB83030EA 0 CNY
KIMB83030 KIMB05812
Wipes for general applications, WYPALL* L30
擦拭巾 一次性湿巾和毛巾 Wipes for General Applications
WypAll® Towels L30 features the double re-creping (DRC) technology, which gives them high surface softness and a reinforced grid for strength. They are designed with the DRC (double re-creping) process, which gives you high surface softness (a towel-like feel) with a reinforced grid that's strong for wet surface wiping and cleaning.
  • Quarter-folded for easy storage
  • Great choice to replace air laid towels
  • Tough enough for light-duty processing, janitorial tasks and maintenance
  • Soft enough for personal use on hands and skin
  • Strong enough for wet surface wiping and cleaning
Ideal for a wide range of light-duty maintenance, processing and janitorial wiping tasks.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904