碳酸氢钠 kit 99,0-100,5 Dried, powder USP, FCC
供应商: Avantor
Synonyms:
Sodium bicarbonate
M.O.C. Category R
BAKR3509-K1EA
0
CNY
BAKR3509-K1
BAKR3509-K3
BAKR3509-K4
BAKR3509-K5
碳酸氢钠 kit 99,0-100,5 Dried, powder USP, FCC
碳酸氢钠
Caution: Not for use in hemodialysis.
分子式:
NaHCO₃ 分子量: 84.01 g/mol 溶点: 270 °C 密度: 2.20 g/cm³ 存储条件: Ambient |
MDL编号:
MFCD00003528 CAS号: 144-55-8 EINECS: 205-633-8 Merck Index: 13,08655 |
产品技术规格
Meets F.C.C. Requirements | |
Meets U.S.P Requirement | |
GMP Manufactured Product | |
Food GMP Manufactured Product | |
Bulk Food Chemical | |
Bulk Pharmaceutical Chemica | |
Kit Components Meet Requirements | Passes Test |