碳酸钠, anhydrous 99.5-100.5% (以无水为基础), granular NF, FCC, J.T.Baker®

供应商: Avantor
Warning


Management of Change (MOC) category = R

BAKRLP08-10EA 0 CNY
BAKRLP08-10 BAKRLP08-S1
碳酸钠, anhydrous 99.5-100.5% (以无水为基础), granular NF, FCC, J.T.Baker®
碳酸钠
分子式: Na₂CO₃
MDL编号: MFCD00003494
CAS号: 497-19-8

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR