氯化钾 饱和水溶液, StandARd®, Macron Fine Chemicals™
供应商: Avantor Performance Materials
Management of Change (MOC) category = N
MACRH300-01EA 422 CNY
MACRH300-01 MACRH300-05
氯化钾 饱和水溶液, StandARd®, Macron Fine Chemicals™
氯化钾
For refilling calomel reference electrodes.
分子式: KCl
密度: 1,13 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00011360
CAS号: 7447-40-7
EINECS: 231-211-8
现在订购
产品技术规格
For Laboratory,Research,or Manufacturing Use
Density (g/mL) at 20°C 1.162 - 1.172
Identification Passes Test