2,2,4-三甲基戊烷 ≥99%, Macron Fine Chemicals™

供应商: Avantor Performance Materials
Danger

Synonyms: 异辛烷

Management of Change (MOC) category = N

MACR6055-19EA 7287 CNY
MACR6055-19
2,2,4-三甲基戊烷 ≥99%, Macron Fine Chemicals™
2,2,4-三甲基戊烷
分子式: (CH₃)₂CHCH₂C(CH₃)₃
分子量: 114.23 g/mol
沸点: 99.2 °C (1013 hPa)
溶点: –107 °C
密度: 0.692 g/cm³ (20 °C)
闪点: –12 °C
存储条件: Ambient
MDL编号: MFCD00008943
CAS号: 540-84-1
EINECS: 208-759-1
UN号: 1262
ADR: 3 II
Merck Index: 13,05212

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use
Specific Gravity at 25°/25°C, For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR