苄醇 98.0-100.5% NF, Multi-Compendial, J.T.Baker®
供应商: Avantor
|
Warning
|
Synonyms:
苯甲醇
Management of Change (MOC) category = R
BAKRBS06-20EA
36156
CNY
BAKRBS06-20
BAKRBS06-50
BAKRBS06-SA
苄醇 98.0-100.5% NF, Multi-Compendial, J.T.Baker®
苄醇
分子式:
C₆H₅CH₂OH 分子量: 108.14 g/mol 沸点: 205 溶点: –15.4…–15.2 °C 闪点: 100.4° C |
MDL编号:
MFCD00004599 CAS号: 100-51-6 Merck Index: 13,01126 |
产品技术规格
GMP Manufactured Product | |
Meets N.F. Requirements | |
Meets B.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Assay (C₆H₅CH₂OH) | 98.0 - 100.5 % |
NF - Clarity of Solution | Passes Test |
NF - Color of Solution | Passes Test |
NF - Identification | Passes Test |
NF - Peroxide Value | ≤ 5 |
NF - Benzaldehyde and Other Related Substances - Benzaldehyde | ≤ 0.15 % |
NF - Related Substances - Cyclohexylmethanol | ≤ 0.10 % |
NF - Benzaldehyde and Other Related Substances - Total Other Peaks | |
of Benzyl Alcohol | ≤ 0.04 % |
Greater Than RT of Benzyl Alcohol | ≤ 0.3 % |
NF - Benzaldehyde and Other Related Substances - Total Other Peaks | ≤ 0.3 % |
NF - Refractive Index, n²ºD | 1.538 - 1.541 |
NF - Acidity | Passes Test |
NF - Residue on Evaporation | ≤ 0.05 % |
NF - Benzene | ≤ 2 ppm |
EP/BP - Assay (C₆H₅CH₂OH) | 98.0 - 100.5 % |
EP/BP - Identification | Passes Test |
EP/BP - Acidity | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Refractive Index, n²º^D | 1.538 - 1.541 |
EP/BP - Benzaldehyde and Other Related Substances - Benzaldehyde | ≤ 0.15 % |
EP/BP - Related Substances - Cyclohexylmethanol | ≤ 0.10 % |
EP/BP - Benzaldehyde and Other Related Substances - Total Other Peaks | ≤ 0.04 % |
EP/BP - Benzaldehyde and Other Related Substances - TOTAL OTHER PEAKS | ≤ 0.3 % |
>RT OF BENZYL ALCOHOL | ≤ 0.3 % |
EP/BP - Peroxide Value | ≤ 5 |
EP/BP - Residue on Evaporation | ≤ 0.05 % |
This product is not intended for use in preparing parenteral dosage | |
forms. | |
The Class 1 solvent Benzene is likely to be present below 2 ppm. The | |
actual analysis of Benzene is reported for each lot. | |
The Class 2 solvent Toluene is likely to be present and is process | |
controlled to <30 ppm. | |
No other Class 1, 2 or 3 residual solvents are likely to be present. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...