碳酸钠, anhydrous 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
供应商: Avantor
|
Warning
|
Synonyms:
纯碱
Management of Change (MOC) category = R
BAKRBS18-10EA
0
CNY
BAKRBS18-10
BAKRBS18-D1
BAKRBS18-D2
BAKRBS18-ST
碳酸钠, anhydrous 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
碳酸钠
分子式:
Na₂CO₃ 分子量: 105.99 g/mol 沸点: 1600 溶点: 854 |
MDL编号:
MFCD00003494 CAS号: 497-19-8 Merck Index: 14,08596 |
产品技术规格
Meets B.P. Chemical Specifications | |
Meets J.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
Meets N.F. Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Water (H₂O) | ≤ 0.5 % |
NF - Assay (Na₂CO₃) (anhydrous basis) | 99.5 - 100.5 % |
EP/BP - Assay (Na₂CO₃) (dried basis) | 99.5 - 100.5 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Identification C | Passes Test |
EP/BP - Identification D | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Alkali Hydroxides and Bicarbonates | Passes Test |
EP/BP - Chloride (Cl) | ≤ 125 ppm |
EP/BP - Sulfate (SO₄) | ≤ 250 ppm |
EP/BP - Iron (Fe) | ≤ 50 ppm |
EP/BP - Loss on Drying | ≤ 1.0 % |
JP - Identification A | Passes Test |
JP - Identification B | Passes Test |
JP - Clarity and Color of Solution | Passes Test |
JP - Chloride (Cl) | ≤ 0.071 % |
JP - Heavy Metals (as Pb) | ≤ 20 ppm |
JP - Arsenic (As) | ≤ 3.1 ppm |
JP - Loss on Drying | ≤ 0.5 % |
JP - Assay (Na₂CO₃) (dried basis) | 99.0 - 101.0 % |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...