Hexanes : acetone mixture 50 : 50 (v/v), ULTRA RESI-ANALYZED®, J.T.Baker®

供应商: Avantor
Danger


Management of Change (MOC) category = NI

BAKR9525-3PEA 0 CNY
BAKR9525-3P BAKR9525-98 BAKR9525-Q3 BAKR9525-Y6
Hexanes : acetone mixture 50 : 50 (v/v), ULTRA RESI-ANALYZED®, J.T.Baker®
Hexanes : acetone mixture
闪点: –23 °C
存储条件: Ambient
UN号: 1993
ADR: 3 II

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use
Acetone - Assay (by GC, corrected for water) ≥99.4 %
Acetone - Color (APHA) ≤10
Acetone - Residue after Evaporation ≤1 ppm
Acetone - Substances Reducing Permanganate Passes Test
Acetone - Titrable Acid (µeq/g) ≤0.3
Acetone - Titrable Base (µeq/g) ≤0.60
Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
Acetone - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
Acetone - GC-ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (ng/L) ≤10
Hexanes - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
Hexanes - GC-ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (ng/L) ≤10
Hexanes - ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL) ≤5
Hexanes- Assay (Total Saturated C₆ Isomers) (by GC, corrected for water) ≥99.5 %
Hexanes- Assay (as n-hexane) (by GC) ≥95 %
Hexanes - Color (APHA) ≤10
Hexanes - Residue after Evaporation ≤1 ppm
Hexanes - Substances Darkened by H₂SO₄ Passes Test
Hexanes - Water (by KF, coulometric) ≤0.05 %
ECD-Sensitive Impurities (as Ethylene Dibromide)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR