甲醇 60% 在水里, BAKER ANALYZED®, J.T.Baker®
供应商: Avantor
|
Danger
|
Management of Change (MOC) category = N
9146-03
9146-07
BAKR9146-03EA
738
CNY
BAKR9146-03
BAKR9146-07
甲醇 60% 在水里, BAKER ANALYZED®, J.T.Baker®
甲醇
Premixed methanol (60%) solution.
分子式:
CH₃OH 密度: 0,90 g/cm³ |
MDL编号:
MFCD00004595 CAS号: 67-56-1 UN号: 1230 ADR: 3 (6.1),II |
产品技术规格
For Laboratory,Research,or Manufacturing Use | |
Assay (CH₃OH) | 58 - 62 % |
Fluorescence | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...