盐酸 37,0 - 38,0%, Finyte®, J.T.Baker®
供应商: Avantor
|
Danger
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
Management of Change (MOC) category = R
BAKR5814-52EA
0
CNY
BAKR5814-52
BAKR5814-54
BAKR5814-58
盐酸 37,0 - 38,0%, Finyte®, J.T.Baker®
盐酸
分子式:
HCl 分子量: 36.46 g/mol 溶点: –30 °C 存储条件: Ambient |
MDL编号:
MFCD00011324 CAS号: 7647-01-0 UN号: 1789 ADR: 8 II |
产品技术规格
For Microelectronic Use | |
Store below 25°C | |
Assay (as HCl) (by acid-base titrn) | 37.0 - 38.0 % |
Color (APHA) | ≤ 5 |
Extractable Organic Substances | ≤ 3 ppm |
Free Halogen (as Cl₂) | Passes Test |
Phosphate (PO₄) | ≤ 0.03 ppm |
Sulfate (SO₄) | ≤ 0.3 ppm |
Sulfite (SO₃) | ≤ 0.4 ppm |
Trace Impurities - Aluminum (Al) | ≤ 5.700 ppb |
Arsenic and Antimony (as As) | ≤ 3.0 ppb |
Trace Impurities - Barium (Ba) | ≤ 1.300 ppb |
Trace Impurities - Beryllium (Be) | ≤ 1.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤ 5.0 ppb |
Trace Impurities - Boron (B) | ≤ 9.200 ppb |
Trace Impurities - Cadmium (Cd) | ≤ 1.0 ppb |
Trace Impurities - Calcium (Ca) | ≤ 23.200 ppb |
Trace Impurities - Chromium (Cr) | ≤ 2.3 ppb |
Trace Impurities - Cobalt (Co) | ≤ 1.300 ppb |
Trace Impurities - Copper (Cu) | ≤ 1.300 ppb |
Trace Impurities - Gallium (Ga) | ≤ 1.0 ppb |
Trace Impurities - Germanium (Ge) | ≤ 5.0 ppb |
Trace Impurities - Gold (Au) | ≤ 5.0 ppb |
Heavy Metals (as Pb) | ≤ 50.0 ppb |
Trace Impurities - Iron (Fe) | ≤ 7.770 ppb |
Trace Impurities - Lead (Pb) | ≤ 1.200 ppb |
Trace Impurities - Lithium (Li) | ≤ 1.300 ppb |
Trace Impurities - Magnesium (Mg) | ≤ 6.000 ppb |
Trace Impurities - Manganese (Mn) | ≤ 1.300 ppb |
Trace Impurities - Molybdenum (Mo) | ≤ 5.0 ppb |
Trace Impurities - Nickel (Ni) | ≤ 1.300 ppb |
Trace Impurities - Niobium (Nb) | ≤ 1.0 ppb |
Trace Impurities - Potassium (K) | ≤ 3.900 ppb |
Trace Impurities - Silicon (Si) | ≤ 30.0 ppb |
Trace Impurities - Silver (Ag) | ≤ 1.0 ppb |
Trace Impurities - Sodium (Na) | ≤ 6.000 ppb |
Trace Impurities - Strontium (Sr) | ≤ 1.0 ppb |
Trace Impurities - Tantalum (Ta) | ≤ 5.0 ppb |
Trace Impurities - Thallium (Tl) | ≤ 5.0 ppb |
Trace Impurities - Tin (Sn) | ≤ 1.820 ppb |
Trace Impurities - Titanium (Ti) | ≤ 1.300 ppb |
Trace Impurities - Vanadium (V) | ≤ 1.0 ppb |
Trace Impurities - Zinc (Zn) | ≤ 4.900 ppb |
Trace Impurities - Zirconium (Zr) | ≤ 10.0 ppb |
Particle Count at point of fill - 0.2 µm and greater | ≤ 2000 par/ml |
Particle Count at point of fill - 0.5 µm and greater | ≤ 100 par/ml |
Particle Count at point of fill - 1.0 µm and greater | ≤ 25 par/ml |
Particle Count at point of fill - 0.1 µm and greater (Rion KS40AF) | ≤ 1680 par/ml |
Particle Count at point of fill - 0.2 µm and greater (Rion KS40AF), For Information Only | |
Particle Count at point of fill - 0.3 µm and greater (Rion KS40AF), For Information Only | |
Particle Count at point of fill - 0.5 µm and greater (Rion KS40AF), For Information Only | |
Particle Count, finished unit - 0.1 µm and greater(Rion KS40AF) (FUT) | ≤ 5050 par/ml |
Particle Count, finished unit - 0.2 µm and greater (Rion KS40AF) (FUT), For Information Only | |
Particle Count, finished unit - 0.3 µm and greater (Rion KS40AF) (FUT), For Information Only | |
Particle Count, finished unit - 0.5 µm and greater (Rion KS40AF) (FUT), For Information Only |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...