Ultra-Low temperature freezers
供应商: IBS STOCK ITEMS
N123001 N113001
IBSAN123001EA 0 CNY
IBSAN123001 IBSAN113001
Ultra-Low temperature freezers
冷柜 超低温冰箱
These ULT freezers are ideal personal easy access freezers, for daily use in laboratories, hospitals or any scientific research area that requires storage of sensitive samples, such as bacteria, viruses, tissue samples or cell preparations.
  • Upright model has temperature logger with USB port and door alarm
  • Chest model has castors, handle lock, audible and visual alarm
  • Both models have high/low temperature alarm
  • Battery backup (50 hours on chest model)
  • ULT C75 model is prepared for CO2 backup
Both the upright and chest freezer need very little maintenance or on-site service, and have low power consumption. The chest freezer offers good stability, reliability, and long-term durability.
Installation is quite easy as these freezers can fit through all standard doors. This combined with their low weight means it is easy install and move either model around.