4-羟基-4-甲基-2-戊酮, J.T.Baker®

供应商: Avantor
Danger

Synonyms: 双丙酮醇

Management of Change (MOC) category = N

BAKRN679-09EA 0 CNY
BAKRN679-09
4-羟基-4-甲基-2-戊酮, J.T.Baker®
4-羟基-4-甲基-2-戊酮
分子式: (CH₃)₂C(OH)CH₂COCH₃
分子量: 116.16 g/mol
沸点: 166 °C (1013 hPa)
溶点: –44 °C
密度: 0.94 g/cm³ (20 °C)
闪点: 58 °C
存储条件: Ambient
MDL编号: MFCD00004471
CAS号: 123-42-2
EINECS: 204-626-7
UN号: 1148
ADR: 3 III
REACH: 01-2119473975-21
Merck Index: 13,02983

现在订购

产品技术规格

For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR