Blotting membranes, Amersham™ Hybond™ ECL™
供应商: GE HEALTHCARE
RPN82D RPN132D
GEHERPN82DEA 0 CNY
GEHERPN82D GEHERPN132D
Blotting membranes, Amersham™ Hybond™ ECL™
转移膜
Hybond™ ECL™ gives excellent sensitivity, resolution, and low background for all labelling and detection systems (radioactive, non radioactive, and chromogenic), especially when used with ECL™, ECL™ Prime, and ECL™ Plex Western Blotting Detection kits and reagents. Special packaging features include interleaf on both sides of the membrane and air-tight resealable aluminium bag with label space for marking the 'opened on' date.
  • 100% pure nitrocellulose membrane validated for use with all ECL™ Western blotting systems and for all protein blotting applications
  • Special packaging features enhance membrane performance in terms of handling and consistency of results
Pore size: 0,45 µm
Minimal protein size required: 7 kDa