Blotting membranes, Amersham™ Hybond™-N+
供应商: GE HEALTHCARE
RPN119B RPN1210B RPN1510B RPN1520B RPN2020B RPN203B RPN303B RPN82B
GEHERPN119BEA 0 CNY
GEHERPN119B GEHERPN1210B GEHERPN1510B GEHERPN1520B GEHERPN2020B GEHERPN203B GEHERPN303B GEHERPN82B
Blotting membranes, Amersham™ Hybond™-N+
转移膜
Amersham™ Hybond™-N+ is a positively charged nylon membrane with a binding capacity for nucleic acids up to 600 µg/cm² recommended for use with radioactive or non radioactive chemiluminescence and chemifluorescence detection systems.
  • Recommended for all high density membrane arrays
  • Multiple reprobing is possible without loss of membrane integrity
  • QC tested in genomic Southern blots
Amersham™ Hybond™-N+ yields excellent sensitivity in conventional Southern blotting. Nucleic acid samples may be fixed by UV exposure or baking, although UV fixation is best for maximum reproducibility.
Pore size: 0,45 µm
Nucleic acid size required: >50 bp
*designed to fit Nunc™ OmniTray.
Caution: Amersham™ Hybond™-N+ is not suitable for alkali blotting and fixation.