试管用冷却盒
供应商: VWR Collection
VWRI479-0641EA 1992 CNY
VWRI479-0641 VWRI479-0642 VWRI479-0643
试管用冷却盒
箱子 冷却/冷冻盒/袋
−20 °C迷你PC试管用冷却盒
,无毒凝胶
−20 °C迷你试管用冷却盒可容纳12×0,5或1,5 ml离心管,并可置于低于−20 °C的温度下长达1小时。适用于工作台或运输用。无毒凝胶填充。使用前,需置于−25 °C冷却器中至少24个小时。不用时,应存储在−25 °C条件下。
Maxi labtop-kylbox för –20 °C, av polykarbonat/icke-toxisk gel−20 °C大号试管用冷却盒可容纳32×0,5或1,5 ml离心管,并可置于低于−20 °C的温度下长达1小时。适用于工作台或运输用。钢丝手柄确保搬运时的稳定性。冷却盒和盖内填充无毒凝胶。不用时,应存储在−25 °C条件下
−20 °C PC试管用冷却盒,无毒凝胶−20 °C试管用冷却盒可以将温度维持在−20 °C和−15 °C之间长达4个小时。可容纳20× 1,5或2,0 ml离心管或冷冻管。含固定0,2到0,5 ml离心管的插入件。印制网格、带键盖子、锁紧手柄、防滑脚垫。不用时,应存储在−25 °C条件下。
The −20 °C labtop cooler maintains temperatures between −20 and −15 °C for up to four hours. Polycarbonate. Holds twenty 1,5 to 2,0 ml microcentrifuge tubes or cryovials. Inserts included hold 0,2 to 0,5 ml microcentrifuge tubes. Printed grid, keyed lid, locking handle, non skid feet. Store at −25 °C between use.