Avantor® Hichrom HI-DEX DET Gamma, GC Columns
供应商: Avantor
HIGCHI69-25-015-15EA
9783
CNY
HIGCHI69-25-015-15
HIGCHI69-25-015-25
HIGCHI69-25-015-30
HIGCHI69-25-015-50
HIGCHI69-25-015-60
HIGCHI69-25-025-15
HIGCHI69-25-025-25
HIGCHI69-25-025-30
HIGCHI69-25-025-50
HIGCHI69-25-025-60
HIGCHI81-10-010-10
HIGCHI81-10-010-5
Avantor® Hichrom HI-DEX DET Gamma, GC Columns
色谱柱
规格
Phase name | HI-DEX DET Gamma |
Environmentally Preferable | |
Phase composition | Diethyl TBS Gamma Cyclodexstrin based |
Polarity | Chiral stationary phase |
Crossbond | No |
温度限值 | 230 °C |