Avantor® Hichrom HI-35 MS, GC Columns
供应商: Avantor
HIGCHI212501030G5EA 9052 CNY
HIGCHI212501030G5 HIGCHI212502515G5 HIGCHI212502530G10 HIGCHI212502530G5 HIGCHI212502560G10 HIGCHI212502560G5 HIGCHI212505015G10 HIGCHI212505030G10 HIGCHI212505030G5 HIGCHI213202530G5 HIGCHI213210030G5 HIGCHI215310030G5 HIGCHI21-18-018-10 HIGCHI21-18-018-20 HIGCHI21-18-018-40 HIGCHI21-18-018-50 HIGCHI21-18-018-60 HIGCHI21-18-020-10 HIGCHI21-18-020-20 HIGCHI21-18-020-40 HIGCHI21-18-020-50 HIGCHI21-18-020-60 HIGCHI21-18-036-20 HIGCHI21-18-036-40 HIGCHI21-18-036-60 HIGCHI21-18-040-10 HIGCHI21-18-040-20 HIGCHI21-18-040-40 HIGCHI21-18-040-50 HIGCHI21-18-040-60 HIGCHI21-20-010-10 HIGCHI21-20-010-20 HIGCHI21-20-010-40 HIGCHI21-20-010-50 HIGCHI21-20-010-60 HIGCHI21-20-020-10 HIGCHI21-20-020-20 HIGCHI21-20-020-40 HIGCHI21-20-020-50 HIGCHI21-20-020-60 HIGCHI21-20-033-12 HIGCHI21-20-033-25 HIGCHI21-20-033-30 HIGCHI21-20-033-50 HIGCHI21-20-033-60 HIGCHI21-20-040-10 HIGCHI21-20-040-20 HIGCHI21-20-040-40 HIGCHI21-20-040-50 HIGCHI21-20-040-60 HIGCHI21-20-100-10 HIGCHI21-20-100-20 HIGCHI21-20-100-40 HIGCHI21-20-100-50 HIGCHI21-20-100-60 HIGCHI21-25-010-15 HIGCHI21-25-010-30 HIGCHI21-25-010-50 HIGCHI21-25-010-60 HIGCHI21-25-025-10 HIGCHI2125025105 HIGCHI21-25-025-15 HIGCHI21-25-025-30 HIGCHI21-25-025-50 HIGCHI21-25-025-60 HIGCHI21-25-050-10 HIGCHI2125050105 HIGCHI21-25-050-15 HIGCHI21-25-050-30 HIGCHI21-25-050-50 HIGCHI21-25-050-60 HIGCHI21-25-100-10 HIGCHI2125100105 HIGCHI21-25-100-15 HIGCHI21-25-100-30 HIGCHI21-25-100-50 HIGCHI21-25-100-60 HIGCHI21-32-010-10 HIGCHI2132010105 HIGCHI21-32-010-15 HIGCHI21-32-010-30 HIGCHI21-32-010-50 HIGCHI21-32-010-60 HIGCHI21-32-025-10 HIGCHI2132025105 HIGCHI21-32-025-15 HIGCHI21-32-025-30 HIGCHI21-32-025-50 HIGCHI21-32-025-60 HIGCHI21-32-050-10 HIGCHI2132050105 HIGCHI21-32-050-15 HIGCHI21-32-050-30 HIGCHI21-32-050-50 HIGCHI21-32-050-60 HIGCHI21-32-100-10 HIGCHI2132100105 HIGCHI21-32-100-15 HIGCHI21-32-100-30 HIGCHI21-32-100-50 HIGCHI21-32-100-60 HIGCHI21-32-150-10 HIGCHI2132150105 HIGCHI21-32-150-15 HIGCHI21-32-150-30 HIGCHI21-32-150-50 HIGCHI21-32-150-60 HIGCHI21-53-010-10 HIGCHI21-53-010-15 HIGCHI21-53-010-30 HIGCHI21-53-025-15 HIGCHI21-53-025-30 HIGCHI21-53-025-50 HIGCHI21-53-025-60 HIGCHI21-53-050-10 HIGCHI21-53-050-15 HIGCHI21-53-050-30 HIGCHI21-53-050-50 HIGCHI21-53-050-60 HIGCHI21-53-100-10 HIGCHI21-53-100-15 HIGCHI21-53-100-30 HIGCHI21-53-100-50 HIGCHI21-53-100-60 HIGCHI21-53-100-75 HIGCHI21-53-150-10 HIGCHI21-53-150-15 HIGCHI21-53-150-30 HIGCHI21-53-150-50 HIGCHI21-53-150-60 HIGCHI21-53-150-75 HIGCHI21-53-300-30 HIGCHI21-05-005-2 HIGCHI21050052.5 HIGCHI21-05-005-5 HIGCHI21-10-010-10 HIGCHI21-10-010-15 HIGCHI21-10-010-5 HIGCHI21-10-020-10 HIGCHI21-10-020-15 HIGCHI21-10-020-5 HIGCHI21-10-040-10 HIGCHI21-10-040-15 HIGCHI21-10-040-5 HIGCHI21-15-015-10 HIGCHI21-15-015-15 HIGCHI21-15-015-20 HIGCHI21-15-015-30 HIGCHI21-15-015-5 HIGCHI21-25-020-30
Avantor® Hichrom HI-35 MS, GC Columns
色谱柱
Proprietary low polarity and low bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and also an excellent choice as a confirmation column for semi-volatile environmental analysis.
  • Phase composition: Cross-bonded low polarity proprietary phase
  • Long lifetime
Max. temperature limits may change depending on the film thickness.