三聚氰酸 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 1,3,5-三嗪-2,4,6-三醇, 2,4,6-三羟基-1,3,5-三嗪

181020010 181020250 181022500
ACRO181020010EA 0 CNY
ACRO181020010 ACRO181020250 ACRO181022500
三聚氰酸 98%
三聚氰酸
分子式: C₃H₃N₃O₃
溶点: >360 °C
密度: 2,5 g/cm³ (20 °C)
MDL编号: MFCD00082990
CAS号: 108-80-5
EINECS: 203-618-0
Merck Index: 13,02728

现在订购

产品技术规格

Appearance White to light beige Crystalline powder and/or lumps
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR