3-Chloro-2-nitropyridine ≥98.0% (by GC)
供应商: TCI
|
Warning
|
C1765-1G
C1765-5G
TCIAC1765-1GEA
0
CNY
TCIAC1765-1G
TCIAC1765-5G
3-Chloro-2-nitropyridine ≥98.0% (by GC)
3-Chloro-2-nitropyridine
分子式:
C₅H₃ClN₂O₂ 分子量: 158.54 g/mol |
MDL编号:
MFCD00490110 CAS号: 54231-32-2 |
|
产品技术规格
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 93.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...