1-溴-2-萘酚 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromonaphthalen-2-ol, 1-Bromo-2-hydroxynaphthalene

107160250 107161000
ACRO107160250EA 0 CNY
ACRO107160250 ACRO107161000
1-溴-2-萘酚 98%
1-溴-2-萘酚
分子式: C₁₀H₇BrO
分子量: 223.07 g/mol
沸点: 314
溶点: 77…81 °C
MDL编号: MFCD00003869
CAS号: 573-97-7
EINECS: 209-363-1

现在订购

产品技术规格

Appearance Off-white to beige to brown-purple Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 84 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR