四丙基高钌酸铵

供应商: Thermo Fisher Scientific
Danger

Synonyms: TPAP

378390010 378392500
ACRO378390010EA 0 CNY
ACRO378390010 ACRO378392500
四丙基高钌酸铵
四丙基高钌酸铵
分子式: C₁₂H₂₈NO₄Ru
分子量: 351.43 g/mol
溶点: 160 °C (dec.)
存储条件: Refrigerator
MDL编号: MFCD00074914
CAS号: 114615-82-6
UN号: 1479
ADR: 5.1 II

现在订购

产品技术规格

Appearance Black Powder
Infrared spectrum Conforms
Ruthenium (Ru) ≥27.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR